Learn More
Microsolv Technology Ez Pack Rsa Vial 2Ml Clr/100Pk DFS Item
Supplier:  Microsolv Technology Corporation 9509C1WCPRS
Vial & Cap Kit includes 100 EA 2ml, Snap Top, Clear RSA™ Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Snap Caps with Silicone Rubber / PTFE, ultra-pure Septa, fitted in the Light Blue Caps. RSA Brand Easy Purchase Pack. 1/PK.
Catalog No. NC0483970
Only null left
Explore available promotions