missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EZ-Magna ChIP™ HT96 Chromatin Immunoprecipitation Kit

Catalog No. 1710078
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1710078 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710078 Supplier MilliporeSigma™ Supplier No. 1710078

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Magna ChIP protein A/G magnetic beads
  • HT96 Nuclei Isolation Buffer
  • HT96 ChIP Buffer
  • Low Stringency IP Wash Buffer
  • HT96 ChIP Elution Buffer
  • Proteinase K Solution
  • Protease Inhibitor Cocktail III
  • 10X Glycine
  • 10X PBS
  • 96 Well ChIP Plate
  • 96 Well Thermal Plate
  • Plate Seal
  • Strip Caps
  • Anti-Trimethyl-Histone H3 (Lys4) -positive control antibody
  • Normal Rabbit IgG -negative control antibody
  • GAPDH Primers - PCR positive control

Requires

Magna Grip HT96 Rack 96-well magnetic separation stand

Reliable streamlined approach works for both cells and tissues and can be performed using either multi-channel pipette or automated liquid handling instrument

  • Complete set of materials for up to 96 ChIP reactions
  • Protein A+G bead blend for ChIP with a broader range of antibodies than A or G alone
  • Low Chromatin requirements: 10, 000 to 100, 000 cells per reaction
  • Includes negative and positive control antibodies and control primer set to simplify validation of experimental procedure
  • Optimized streamlined protocol with only a single buffer for sonication, IP, or wash; and protocols for automated liquid handling systems
  • Protocols for using cells or tissues
  • Direct analysis of DNA without additional clean-up steps
  • Compatible with ChIPAb+ validated antibody and primer sets

Specifications

Content And Storage store components at the temperatures indicated on the labels
For Use With (Application) Allows the performance of chromatin Immunoprecipitation in a 96-well plate-based format
Quantity 96 Assays
Stability Kit components are stable for 1 year from date of shipment when stored as directed