missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotinylation Kit

Catalog No. pi21435
Change view
Click to view available options
Quantity:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21435 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21435 Supplier Thermo Scientific™ Supplier No. LSG21435
Only null left

Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Highlights:

  • Water-soluble analog of NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate or phosphate buffers
  • Negative charge of -SO3 group keeps biotinylation localized at the cell surface
  • Creates an amide bond
U.S. Patents 5,892,628 and 5,872,261.

Specifications

Description EZ-Link Sulfo-NHS-LC-Biotinylation Kit
Product Type Sulfo-NHS-LC-Biotinylation Kit
Quantity 10 reactions
Content And Storage Sufficient For: 10 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link Sulfo-NHS-LC-Biotin, 25 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10 mg

Store biotin and avidin reagents at -20°C. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins

For Research Use Only. Not for use in diagnostic procedures.