missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotinylation Kit

Catalog No. pi21425
Change view
Click to view available options
Quantity:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21425 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21425 Supplier Thermo Scientific™ Supplier No. 21425
Only null left

Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.

Highlights:

  • Shorter spacer arm than Sulfo-NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate, or phosphate buffers
  • Negative charge of -SO3 keeps biotinylation localized at the cell surface so reagent does not pass through cell membrane
  • Water solubility eliminates the need for harmful organic solvents
U.S. Patents 5,892,628 and 5,872,261.

Specifications

Description EZ-Link Sulfo-NHS-Biotinylation Kit
Product Type Sulfo-NHS-Biotinylation Kit
Quantity 10 reactions
Content And Storage Sufficient For: 10 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link Sulfo-NHS-Biotin, 25 mg
• PBS Pack (makes 500mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10 mg

Store avidin and vial of biotin reagent (with desiccant) at -20°C. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins

For Research Use Only. Not for use in diagnostic procedures.