Learn More
Air-Tite EZ-Dose Syringe
Built for precision and reliability, EZ-Dose Syringes offer exceptional quality and durability.
$42.28 - $149.34
Specifications
Autoclavable Not Autoclavable
Needle Gauge No Needle Included
Latex Free Yes
Product Type Disposable Syringe
Disposable Single-use
View More Specs
Products 12
Catalog Number Mfr. No. Volume (Metric) Connection Type Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Connection Type Price Quantity  
14-818-151
View Documents
Air-Tite
MEZ1LL
1 mL Centric Luer Lock
Pack of 100 Each for $83.45
Case of 32 Pack for $2,670.40
Only null left
14-818-152
View Documents
Air-Tite
MEZ1LS
1 mL Centric Luer Slip
Pack of 100 Each for $42.28
Case of 32 Pack for $1,352.96
Only null left
14-818-147
View Documents
Air-Tite
MEZ3LL
3 mL Centric Luer Lock
Pack of 100 Each for $42.28
Case of 24 Pack for $1,014.72
Only null left
14-818-148
View Documents
Air-Tite
MEZ3LS
3 mL Centric Luer Slip
Pack of 100 Each for $84.45
Case of 24 Pack for $2,026.80
Only null left
14-818-155
View Documents
Air-Tite
MEZ5LL
5 mL Centric Luer Lock
Pack of 100 Each for $50.52
Case of 16 Pack for $808.32
Only null left
14-818-156
View Documents
Air-Tite
MEZ5LS
5 mL Centric Luer Slip
Pack of 100 Each for $84.45
Case of 16 Pack for $1,351.20
Only null left
14-818-146
View Documents
Air-Tite
MEZ10LL
10 mL Centric Luer Lock
Pack of 100 Each for $66.97
Case of 12 Pack for $803.64
Only null left
14-818-145
View Documents
Air-Tite
MEZ10LS
10 mL Centric Luer Slip
Pack of 100 Each for $66.97
Case of 12 Pack for $803.64
Only null left
14-818-149
View Documents
Air-Tite
MEZ20LL
20 mL Centric Luer Lock
Pack of 100 Each for $87.55
Case of 6 Pack for $525.30
Only null left
14-818-150
View Documents
Air-Tite
MEZ20LS
20 mL Eccentric Luer Slip
Pack of 100 Each for $84.45
Case of 6 Pack for $506.70
Only null left
14-818-153
View Documents
Air-Tite
MEZ50LL
50 to 60 mL Centric Luer Lock
Pack of 100 Each for $149.34
Case of 4 Pack for $597.36
Only null left
14-818-154
View Documents
Air-Tite
MEZ50LS
50 to 60 mL Eccentric Luer Slip
Pack of 100 Each for $84.45
Case of 4 Pack for $337.80
Only null left
Description
Description
Designed exclusively for laboratory and veterinary use, EZ-Dose Syringes come in eco-friendly, fully recyclable packaging, delivering performance you can trust, with sustainability in mind.
Specifications
Specifications
Not Autoclavable
Yes
Single-use
Yes
No Needle Included
Disposable Syringe
Hypodermic Needle, Safety Needle, Blunt Fill Needle
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'