Learn More
Micronova™ Extreme Temperature Polyimide Cleanroom Tape
Extreme temperature polyimide tape is suitable for high heat and sub zero applications (-100°F to 500°F). The pressure-sensitive silicone adhesive ensures a clean release from most substrates, even after extreme temperature exposure.
Supplier:  Micronova™ 82538AM
 View more versions of this product
Catalog No. 191600099
Only null left
Description
Description
The Micronova Extreme Temperature Polyimide Cleanroom Tape features transparent amber polyimide film with pressure-sensitive silicone adhesive. Designed with extreme temperature resistance, this tape is ideal for heat sealing, solder mask for circuit boards, flame resistant electrical insulation, lamination, cold storage, lyophilization and freeze drying.
  • Transparent amber polyimide film with pressure-sensitive silicone adhesive
  • Extreme temperature resistant properties: -100°F to 500°F (-70°C to 260°C)
  • Suitable for high heat and sub zero applications
  • Clean release from most substrates, even after extreme temperature exposure
  • Ideal for flame resistant electrical insulation, solder mask for circuit boards, heat sealing, lamination, cold storage, lyophilization and freeze drying
  • Resistant to most chemicals including hydrocarbons, alcohols, and halogenated solvents
  • Wound on a plastic core
Specifications
Chemical Identifiers
Not Autoclavable
Silicone, Pressure-sensitive
Amber
Polyimide
Non-sterile
MicronovaTM P52W Tape Dispenser
Certificate of Conformance, ISO 9001 Certified
Polymer
Tape
Extreme Temperature Cleanroom Tape
Suitable for high heat and subzero applications and ensures a clean release on Ideal for flame resistant electrical insulation, solder mask for circuit boards, heat sealing, lamination and test applications, over-labeling, subzero storage, lyophilization and freeze drying
Yes
Specifications
Not Autoclavable
Silicone, Pressure-sensitive
Amber
Tape
Extreme Temperature Cleanroom Tape
Suitable for high heat and subzero applications and ensures a clean release on Ideal for flame resistant electrical insulation, solder mask for circuit boards, heat sealing, lamination and test applications, over-labeling, subzero storage, lyophilization and freeze drying
Yes
-100°F to +500°F
36 yd. x 0.375 in.
33 m
9.525 mm
9.525 mm
Certificate of Conformance, ISO 9001 Certified
Polymer
High Gloss
Polyimide
Non-sterile
MicronovaTM P52W Tape Dispenser
48/Cs.
-70°C to +260°C
36 yd.
0.375 in.
0.375 in.
Product Suggestions
Product Suggestions