missing translation for 'onlineSavingsMsg'
Learn More

Watson Gloves Extreme Gloves Anti Vibe

Catalog No. 17099871
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17099871 Small
17099895 Medium
17099884 Large
17099818 X-Large
17099881 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17099871 Supplier Watson Gloves Supplier No. 010BKS
Only null left

Extreme Gloves Anti Vibe have heavyweight Durafibre™ microfibre palm with D3O™ Zero™ foam padded palm patches for added protection. WasteNot™ Spandex back with Camo pattern provides form fitty memory.

  • Heavyweight Durafibre™ microfibre palm with recycled grade D3O™ Zero™ foam padded palm patches
  • Spandex back made with WasteNot™ polyester yarn which is comprised of 96% recycled PET bottles
  • Polyurethane printed fingertip and thumb is compatible with all touchscreen devices
  • Heavy duty rubber on back of hand and fingers
  • Snug-fitting neoprene wrist with secure Velcro™ closure

Specifications

Applications Industrial
Coating Material Uncoated
Color Black
Disposable Reusable
Exterior Finish Partially Textured
Hand Ambidextrous
Latex Free Yes
Material Microfiber
Product Type Work Gloves
Size Small
Sterility Non-sterile
Style Gunn
For Use With (Application) Partially Textured
For Use With (Equipment) No
Liner Material Yes
Palm Type Microfiber
Temperature (English) Min Small
Temperature (Metric) Min. Gunn
Show More Show Less