missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Extran™ AP 18

Use for everyday cleaning of laboratory glassware and equipment. MilliporeSigma Extran™ AP 18 is a mildly alkaline concentrated liquid cleaning agent for automatic washers. Extran products are biodegradable, with no residues or NTA. (Note: this produ

$226.48 - $1382.70

Specifications

Form Concentrated Liquid
pH 13.5 (aq. soln., 20°C)
Appearance Colorless to Light Yellow
Composition Aqueous solution of inorganic and organic compounds.
For Use With (Application) Automatic washing
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Container Type Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Container Type Price Quantity  
M1401182500
View Documents
MilliporeSigma™
1401182500
2.5 L Plastic bottle
Each for $226.48
Only null left
 
M1401185000
View Documents
MilliporeSigma™
1401185000
5 L Plastic bottle
Each for $386.14
Only null left
 
M1401189010
View Documents
MilliporeSigma™
1401189010
10 L Plastic container
Each for $688.51
Only null left
 
M1401189025
View Documents
MilliporeSigma™
1401189025
25 L Plastic container
Each for $1,382.70
Only null left
 
Description

Description

Specifications

Specifications

Concentrated Liquid
Colorless to Light Yellow
Automatic washing
13.5 (aq. soln., 20°C)
Aqueous solution of inorganic and organic compounds.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'