Learn More
MilliporeSigma™ Extran™ AP 11
Use for everyday cleaning of laboratory glassware and equipment. MilliporeSigma Extran™ AP 11 is a mildly alkaline powdered cleaning agent for automatic washers. Extran products are biodegradable, with no residues, impurities or harsh chemicals. (Not
Supplier:  MilliporeSigma™ 1.07558.9010
 View more versions of this product
Catalog No. M1075589010
Only null left
Description
Description
A multi-purpose powdered detergent for heavy-duty laboratory cleaning applications. extran™ MN01 is a mixture of anionic surfactants and auxiliary agents. Can be used on glass, plastic, rubber, metal and quartz. Articles to be cleaned should be immersed in a cleaning solution and thoroughly brushed or wiped. Rinsing with distilled water will leave a residue-free surface.
Specifications
Chemical Identifiers
Powdered Solid
White
Automatic washing
11.3 (3 g/L aq. soln., 20°C)
700 kg/m3
Specifications
Powdered Solid
White
700 kg/m3
10 kg
11.3 (3 g/L aq. soln., 20°C)
Plastic bucket
Automatic washing
Product Suggestions
Product Suggestions