missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Extended-Length Tips
GREENER_CHOICE

Catalog No. 02681421
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Lift-off Rack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
02681421 100 to 1000 μL Non-sterile Bulk
02681419 1 to 200 μL Non-sterile Lift-off Rack
02681420 1 to 200 μL Sterile Lift-off Rack
02681418 1 to 200 μL Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02681421
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extended length tips are ideal for difficult-to-reach samples

  • Easily reach the bottom of long tubes
  • Minimize pipet contamination by preventing pipet shaft from contacting the tube or vessel
  • Natural color
  • Certified by independent laboratory– Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) 3.6 in.
Length (Metric) 9cm
Clean Claims DNase, RNase, DNA, Bioburden, and PCR inhibitor Free
Format Bulk
Tip Style Universal
Packaging BP
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less