missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Extended-Length Tips
GREENER_CHOICE

Catalog No. 02681419
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Lift-off Rack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
02681419 1 to 200 μL Non-sterile Lift-off Rack
02681421 100 to 1000 μL Non-sterile Bulk
02681420 1 to 200 μL Sterile Lift-off Rack
02681418 1 to 200 μL Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02681419

Extended length tips are ideal for difficult-to-reach samples

  • Easily reach the bottom of long tubes
  • Minimize pipet contamination by preventing pipet shaft from contacting the tube or vessel
  • Natural color
  • Certified by independent laboratory– Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) 3.6 in.
Length (Metric) 91 mm
Clean Claims DNase, RNase, DNA, Bioburden, and PCR inhibitor Free
Format Lift-off Rack
Tip Style Universal
Packaging LR, 8 x 204
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less