missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Expression System Kit

Catalog No. a40003643
Change view
Click to view available options
Quantity:
1 Starter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40003643 1 Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40003643 Supplier Gibco™ Supplier No. A40003643
Only null left

The Expi293 PRO Expression System enables fast, high-yield protein production from HEK293 cells. Designed for efficiency and scalability, it delivers superior expression of complex proteins with a simplified, automation-friendly workflow.

The Gibco Expi293 PRO Expression System is a complete solution for rapid, high-yield protein production from HEK293 cells. It is a fully integrated expression system designed to deliver higher yields of a wider variety of proteins—including challenging and low-yield proteins—with exceptional speed, efficiency, and an easy-to-automate protocol. Producing increasingly complex proteins with higher yields and increased throughput is critical to accelerating breakthroughs in protein research applications and drug development.

The Expi293 PRO Expression System features include:

  • Unlock new protein targets; express challenging and low-yield proteins
  • Exceptional protein yields; multi-fold increase in protein yields—up to 5 g/L
  • Cost effective; the lowest cost/mg
  • Rapid production of recombinant proteins in as little as 2–4 days
  • High throughput and automation compatible
  • High density cultures of Expi293 PRO cells that result in more expressing cells per milliliter of culture
  • Reduce reagents, plasticware, and labor by 3X
  • Native folding and mammalian post-translational modification of expressed proteins
  • Scalable expression from <1 mL to >10 L

The components included in the Expi293 PRO Expression System Kit are: two vials of frozen Expi293 PRO cells, one liter of Expi293 PRO Expression Medium, one Expi293 PRO Transfection Kit sufficient to transfect 1 liter of culture, Opti-Plex Complexation Buffer, and an antibody-expressing positive control vector. All components of the Expi293 PRO Expression System are also available for individual purchase.

Specifications

Cell Line Expi293 PRO
Cell Type HEK293
Format Kit
Promoter pcDNA™ 3.4
Transfection Technique Lipid-based Transfection, Transient Transfection
Species Human
Product Type Expression System Kit
Quantity 1 Starter Kit
Sterility Sterile

For Research Use Only. Not for use in diagnostic procedures.