Learn More
Gibco™ Expi293™ PRO Expression Medium
Description
Gibco Expi293 PRO Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293 PRO cells in the Expi293 PRO Expression System for scalable transient protein expression.
- Supports growth of suspension HEK 293 cultures to densities of over 15 x 10^6 cells/mL
- Transfection-compatible serum-free suspension culture medium
- Helps enable sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 5 g/L of recombinant protein
- Supports growth and transfection of HEK293 cells in culture formats of 1 mL to 10 L
- Does not contain phenol red
Expi293 PRO Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 PRO Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.
Specifications
Specifications
| Cell Type | Embryonic Kidney Cells |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | Expression Medium |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Sterilization Method | Sterile-filtered |
| With Additives | GlutaMAX |
| Without Additives | No HEPES, No Phenol Red |
| Product Line | Expi293 PRO |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.