Learn More
Gilson™ EXPERTTM Universal Fit Standard Pipette Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
Supplier:  Gilson™ F1736011
 View more versions of this product
Catalog No. F1736011G
Only null left
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
Standard Pipette Tips
Non-filtered
Clear
E300ST Sterilized Tipack
RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
Rack
Racked
Air Displacement Tips
Single-use
Yes
Polypropylene
10, 50, 100μL
1 to 300 μL
Sterile
Autoclavable
5.857cm
Pipetting
Standard
768/Cs.
Green
Universal Fit
EXPERT
Conical
Product Suggestions
Product Suggestions