missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ EXPERTTM Standard TOWERPACKTM Reload System Pipette Tips
Description
- 10 layers of tips snap with ease into Tipack boxes
- Clear packaging allows easy identification of tips
- Color-coded tip packaging for volume color matching to pipettes
- Graduations tips enable quick-checks when aspirating samples
- Economical and easy-to-use
- Manufactured in sterile, clean-room environment
- Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
| Description | Standard TowerPack Reload System Pipette Tips |
| Volume (Metric) | 1 to 300 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Model | E300 |
| Length (Metric) | 5.857cm |
| Clean Claims | RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free |
| For Use With (Application) | Pipetting |
| Show More |