missing translation for 'onlineSavingsMsg'
Learn More

exo-Tetrahydrodicyclopentadiene 94.0+%, TCI America™

Catalog No. t19945g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19945G 5 g
T199425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19945G Supplier TCI America Supplier No. T19945G

Chemical Identifiers

CAS 2825-82-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD01714931
InChI Key LPSXSORODABQKT-UHFFFAOYNA-N
Synonym exo-Tricyclo[5.2.1.0(2,6)]decane
PubChem CID 11159354
IUPAC Name tricyclo[5.2.1.0²,⁶]decane
SMILES C1CC2C3CCC(C3)C2C1

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 136.24
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material exo-Tetrahydrodicyclopentadiene
EINECSNumber (4)-0655
RTECSNumber PB9600000
TSCA Yes