Learn More
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 98%
CAS: 29745-04-8 | C8H8O4 | 168.148 g/mol
Supplier: Thermo Scientific Chemicals L0294714
Description
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsexo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with Strong acids, Strong bases.
Specifications
| exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride | |
| 111°C to 116°C | |
| MFCD00213361 | |
| 84287 | |
| exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin | |
| JAABVEXCGCXWRR-GUCUJZIJSA-N | |
| 168.148 | |
| 168.15 |
| 29745-04-8 | |
| C8H8O4 | |
| 25 g | |
| Moisture sensitive | |
| Hydrolyzes in water. | |
| C1CC2C3C(C1O2)C(=O)OC3=O | |
| 12251906 | |
| ≥98% |
Chemical Identifiers
| 29745-04-8 | |
| 168.148 | |
| JAABVEXCGCXWRR-GUCUJZIJSA-N | |
| 12251906 |
| C8H8O4 | |
| MFCD00213361 | |
| exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin | |
| C1CC2C3C(C1O2)C(=O)OC3=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
RTECSNumber : RN8300000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only