missing translation for 'onlineSavingsMsg'
Learn More

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 98%

Catalog No. aal0294706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0294706 5 g
AAL0294714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0294706 Supplier Thermo Scientific Chemicals Supplier No. L0294706
Only null left

CAS: 29745-04-8 | C8H8O4 | 168.148 g/mol

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with Strong acids, Strong bases.

Chemical Identifiers

CAS 29745-04-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00213361
InChI Key JAABVEXCGCXWRR-GUCUJZIJSA-N
Synonym exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin
PubChem CID 12251906
SMILES C1CC2C3C(C1O2)C(=O)OC3=O

Specifications

Melting Point 111°C to 116°C
Quantity 5 g
Beilstein 84287
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber RN8300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only