missing translation for 'onlineSavingsMsg'
Learn More
Learn More
exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, 98+%
Description
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
Specifications
| Melting Point | 118°C to 119°C |
| Quantity | 25 g |
| Beilstein | 10355 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 166.13 |
| Percent Purity | ≥98% |
| Chemical Name or Material | exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 226-570-2 |
| RTECSNumber | KC9650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only