missing translation for 'onlineSavingsMsg'
Learn More

exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7128977
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E039225G 25 g
E0392100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E039225G Supplier TCI America Supplier No. E039225G
Only null left

Chemical Identifiers

CAS 6118-51-0
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00151506,MFCD00077442,MFCD00151506,MFCD00151506
InChI Key QQYNRBAAQFZCLF-UHFFFAOYNA-N
Synonym exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
PubChem CID 124671625
IUPAC Name 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1OC(=O)C2C3OC(C=C3)C12

Specifications

Color White
Quantity 25 g
Formula Weight 166.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic Anhydride
RTECSNumber KC9650000
TSCA Yes