Learn More
Welch™ Exhaust Filter with Drainback Feature
Catalog No. 01184114
Change view
Click to view available options
Type:
Exhaust Filter
For Use With:
Welch CRVPro2, 4, 6, 8
Catalog No. Type For Use With
01184114 Exhaust Filter Welch CRVPro2, 4, 6, 8
1 options
Catalog No. 01184114 Supplier Welch™ Supplier No. 1417E
Only null left
Coalesces exhaust oil mist from rotary vane vacuum pump and drains back into oil case to reduce oil consumption and enable a clean work space. Exhaust can be directed into existing fume hood. Filters to 0.3 μm particles.
  • Exhaust can be directed into existing fume hood
  • Reduces pump noise
  • Use 1417E for pumps with NW16 Exhaust Connections (such as Welch CRVPro2, 4, 6, 8).
  • Use 1417F for pumps with NW25 Exhaust Connections (such as Welch CRVpro16, 24, 30).
  • Use 1417H-01 for replacement element for 1417F
Specifications
For Use With (Application) Filters to 0.3 μm particles
For Use With Welch CRVPro2, 4, 6, 8
Size Connection: NW16
Type Exhaust Filter