Learn More
BINDER™ Execution of IQ/OQ
Execution of IQ/OQ in accordance with qualification folder.
Supplier: BINDER™ DL400200
Description
- Installation Qualification (IQ) confirms that the unit has been properly installed according to customer requirements including documentation. The technician checks that the unit has been installed correctly, as specified in the qualification folder. The qualification folders can be ordered on a unit-specific basis.
- OQ (Operational Qualification) checks and confirms that the unit is operating properly in an unloaded state. The required tests are available in the qualification folder.
Specifications
| Environmental Walk-in Chambers | |
| IQ/OQ Service |