Learn More
Excelta™ Roto-Pic™ Vacuum System Probe Kit
Catalog No. 17468534
Change view
Click to view available options
For Use With (Equipment):
Roto-pic vacuum system
Catalog No. For Use With (Equipment)
17468534 Roto-pic vacuum system
1 options
Catalog No. 17468534 Supplier Excelta™ Supplier No. 2000P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Allow the tips to be rotated by operating the 'wheel' on the hand piece.
The 2000P Roto-pic™ vacuum probe kit contains one each of the 2020 (40°), 2021 (60°), 2022 (45° Short tip), 2023 (90° Tip) and 2024 (straight tip).
Specifications
Product Type Vacuum Pickup Roto-pic 5 piece Probe kit
For Use With (Equipment) Roto-pic vacuum system
Includes 40° 2020 tip, 60° 2021 tip, 45° 2023 short tip, 90° 2024 straight tip