missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Excelta™ Precision Tweezers with Straight Strong Medium Tips
Precision Tweezers feature strong tips for long life.
Supplier: Excelta™ 00BSAPI
Description
Strong tips for long life
- Anti-magnetic stainless steel or Carbon Steel
Chemical Identifiers
| Tweezers, Electronic | |
| 0.015 in. | |
| 0.32 in. |
| 1 Ea. | |
| 0.03 cm | |
| 0.8 cm |
Specifications
| Straight Strong, Fine | |
| 4.75 in. | |
| 120 mm | |
| 1 Ea. | |
| 0.03 cm | |
| 0.32 in. | |
| High-precision |
| 00B | |
| Anti-magnetic Stainless Steel | |
| Tweezers, Electronic | |
| 0.015 in. | |
| Fine | |
| 0.8 cm |