Learn More
Excelta™ Precision Tweezers with Straight Strong Medium Tips
Precision Tweezers feature strong tips for long life.
Supplier:  Excelta™ 0SASE
 View more versions of this product
Catalog No. 17447146
Only null left
Description
Description
Strong tips for long life
  • Anti-magnetic stainless steel or Carbon Steel
Specifications
Chemical Identifiers
Tweezers, Electronic
0.015 in.
0.32 in.
1 Ea.
0.03 cm
0.8 cm
Specifications
Strong Straight Serrated Blunt, Precision
4.75 in.
120 mm
1 Ea.
0.03 cm
0.06 in.
0.0067 in.
0.017 cm
0.1 mm
0
Anti-magnetic Stainless Steel
Tweezers, Electronic
0.015 in.
General
0.32 in.
0.8 cm
0.0039 in.
High-precision
Product Suggestions
Product Suggestions