Learn More
Excelta™ Pen-Vac™ Lightweight Vacuum Pens
Description
- High-precision
- Plunger in rear of tool activates vacuum
- With two 0.50 and 0.63 in. (1.3 and 1.6cm) static dissipative Buna-N vacuum cups, that slip onto a barbed fitting
Specifications
Specifications
| Product Type | Lightweight vacuum pen |
| For Use With (Application) | Moving into and out of burn in tray, test socket or other transfer operation |
| Includes | Two 0.5 and 0.63 in. static dissipative Buna-N vacuum cups, that slip onto a barbed fitting |