Learn More
Excelta™ Miniature Tweezers
The shorter length of these tweezers makes them well suited to the limited working area of a microscope
Supplier:  Excelta™ M4AS
 View more versions of this product
Catalog No. 17456070
Only null left
Description
Description
High-precision stainless steel.
Specifications
Chemical Identifiers
Tweezers, Miniature
For microscopy
0.038 cm
0.25 in.
Each
0.015 in.
High
0.63 cm
Specifications
Tapered, Fine
Stainless Steel
Tweezers, Miniature
For microscopy
0.038 cm
0.25 in.
4A
3.5 in.
89 mm
Each
0.015 in.
High
0.63 cm
Product Suggestions
Product Suggestions