missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Excelsior™ ES Tissue Processor SecureSette Basket Kit

Catalog No. A82310071
Change view
Click to view available options
Product Type:
Tissue Processing Basket Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A82310071 Tissue Processing Basket Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A82310071 Supplier Epredia™ Supplier No. A82310071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accommodate the SecureSette tissue cassettes with the Epredia™ Excelsior™ ES SecureSette Basket Kit.

  • New design which accommodates SecureSette tissue processing cassettes
  • Microwaveable design allowing pre-processing of tissue
  • Each kit accommodates 42 SecureSette tissue processing cassettes

Compatible with:

Excelsior™ ES Tissue Processor

Specifications

Product Type Tissue Processing Basket Kit
Content And Storage Ambient