Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Description
Product Features:
- Microwaveable design allowing pre-processing of tissue
- Organized capacity of 300 cassettes in six basket-halves
Ordering Alerts:
Basket halves only match with each other but can be mixed with 222 cassette baskets or SecureSette Excelsior ES baskets
Includes:
Six 50-cassette basket halves and user guide and loading instructions
Compatible with:
Excelsior ES Tissue Processor
Specifications
Specifications
| Product Type | Potassium Permanganate Filter |
| For Use With (Equipment) | Excelsior AS Tissue Processor |
| Quantity | Cs. of 6 Filters |