missing translation for 'onlineSavingsMsg'
Learn More

ACCU-SCOPE™ Excelitas Mini LED Fluorescence Illuminator

Catalog No. 01000942
Change view
Click to view available options
Camera Type:
Nikon
Olympus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Camera Type
01000942 Olympus
01000941 Nikon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01000942 Supplier ACCU-SCOPE™ Supplier No. 01000479R32X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The X-Cite mini™ is the new go-to white-light LED light source for fluorescence imaging applications.

  • Includes: X-Cite mini+ Head, miniCube, speedDIAL
  • Wavelength Range Model: XTMS: 360-700 nm, Model: XTML: 380-700 nm
  • External Power Supply Universal input 100-240 VAC, 50/60 Hz
  • Current 2.4-1.2 A
  • LED ON/OFF Response Times 100μs TTL / 1ms USB
  • Control Options ON/OFF TTL compatible, speedDIAL, RS-232 commands (SDK available), USB
  • I/O Connections BNC input, USB (B-type), 3.5 mm stereo plug
  • Dimensions – HEAD 100 mm x 135 mm x 110 mm (W x H x D)
  • Dimensions – CONTROLLER 180 mm x 110 mm x 230 mm (W x H x D)
  • Dimensions – speedDIAL 80 mm x 59 mm x 112 mm (W x H x D)
  • Weight 3.4 kg (7.5 lb)
  • Certifications RoHS, CE
  • Warranty LEDs – 20,000 hours, All other X-Cite mini+ components – 1 year, parts and labor

Specifications

Type Illuminator
Autoclavable Not Autoclavable
Camera Type Olympus
For Use With (Equipment) Microscope
Voltage 100/240 V