missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ M7000 Imaging System

Catalog No. 12563632
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12563632 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12563632 Supplier Invitrogen™ Supplier No. AMF7000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-performance, fully automated, inverted, multi-channel fluorescence and transmitted light imaging system. Developed to meet today’s expectations for image quality, user interface interactions, data generation speed and flexibility, and end-user value with just a few clicks.

  • Outstanding image quality and versatility with a 5-position objective turret, 4-color LED fluorescence and transmitted light channels, and 3.2 MP CMOS color and B/W cameras
  • Exceptional usability with fully automated X/Y scanning stage, autofocus, and acquisition routines
  • High-speed image acquisition coupled with multi-position well scanning, and Z-stack and tile-stitch options lend power and flexibility to your data generation
  • Fully integrated time-lapse live cell imaging using the optional EVOS Onstage Incubator for precise control of temperature, gases for normoxic or hypoxic conditions, and humidity
  • Powerful image analysis capabilities for cell segmentation and quantification with the optional Celleste™ Image Analysis Software
  • Combines and integrates precision components with a unique modern design functionality that enables high-quality automated fluorescence imaging anywhere with unprecedented workflow efficiency. Full automation of the precision X/Y-stage movement; changing of the 4 LED fluorescent light cubes, 5-position objective turret, focus, and exposure; and switching of the dual camera make the EVOS M7000 system an exceptional platform for a variety of demanding imaging applications
  • Can program to run well-plate scanning, time lapse experiments, and tile-stitch/montage area scans in Z-stack and/or time-lapse modes in your vessel of choice
  • Compatible with all accessories for the predecessor EVOS FL Auto imaging system
  • Versatile and highly configurable with a 5-position objective turret and the ability to simultaneous acquire images in four fluorescence channels plus transmitted light
  • An extensive suite of tools for acquisition, visualization, and analysis
  • Smart LED illumination technology
  • Easy to use and reliable
  • *Note that three (3) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug: AMEP4644 EVOS Power Cord, Type A (North America) AMEP4645 EVOS Power Cord, Type G (United Kingdom) AMEP4646 EVOS Power Cord, Type C (Europe) AMEP4647 EVOS Power Cord, Type I (Australia) AMEP4648 EVOS Power Cord, Type L (Italy) AMEP4649 EVOS Power Cord, Type H (Israel) AMEP4708 EVOS Power Cord, Type D (India/South Africa) AMEP4782 Power Cord, Type I (China, CCC certified) AMEP4786 Power Cord Type J (Switzerland)

    Specifications

    Type Cell Imaging System
    Product Line EVOS
    Camera 3.2 MP Color CMOS
    For Use With (Application) Neurobiology, Immuno-Oncology, Live-Cell Imaging, 3D Cell Imaging, High-Resolution Tile Scanning, Immunohistochemistry (IHC)
    Model M7000
    Light Source LED
    Magnification 1.25X to 100X
    Objectives 5 Position Turret
    Resolution 1920 x 1080 pixels
    Display LCD
    Content And Storage

    EVOS M7000 Imaging System

    • 27-inch 4K LCD monitor with 3840 x 2160 pixel resolution
    • External Dell computer with Intel Core i9 processor, 128 GB DDR5 RAM, 4 TB SSHD, and NVIDIA Quadro RTXA4000 with 16 GB discrete video graphics running Windows 11 LTSC

    EVOS M7000 Accessory Kit

    • EVOS M7000 Quick Start Guide
    • 16 GB USB 3.0 drive with user manual
    • Universal power supply, 24V, 5A
    • 3 North America (Type B) power cords*
    • USB 3.0 type A to B cable
    • Display port to display port cable, 6 ft
    • Cube access tool
    • EVOS calibration slide (AMEP4720)
    • Vessel holder with retention for two slides (AMEPVH021)
    • Vessel holder with retention for microwell plates (AMEPVH022). Also functions as adapter for vessel holders AMEPVH001 through AMEPVH018.
    • Block condenser slider (AMEP4688)
    • Diffuser condenser slider for brightfield applications (AMEPDFS1)
    • Slider for 4X objectives (AMEP4738)
    • EVOS light shield
    • Dust cover
    • Shipping restraint removal guide
    • Mouse pad
    • Tech Support card
    • Corded optical USB mouse
    • Keyboard

    * Note that three (3) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug:

    • AMEP4644 EVOS Power Cord, Type A (North America)
    • AMEP4645 EVOS Power Cord, Type G (United Kingdom)
    • AMEP4646 EVOS Power Cord, Type C (Europe)
    • AMEP4647 EVOS Power Cord, Type I (Australia)
    • AMEP4648 EVOS Power Cord, Type L (Italy)
    • AMEP4649 EVOS Power Cord, Type H (Israel)
    • AMEP4708 EVOS Power Cord, Type D (India/South Africa)
    • AMEP4782 Power Cord, Type I (China, CCC certified)
    • AMEP4786 Power Cord Type J (Switzerland)
    Quantity 1 system
    Shipping Condition Room Temperature
    Show More Show Less
    Can I visualize Alexa Fluor 488 dye using the EVOS M7000 Imaging System?

    Yes, you can visualize Alexa Fluor 488 dye using the EVOS M7000 Imaging System. The system supports the GFP light cube, which is compatible with Alexa Fluor 488 dye. This allows for effective imaging of samples stained with Alexa Fluor 488 dye.

    For more details on compatible dyes and light cubes, please refer to page 126 of the EVOS M5000 Imaging System User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018326_EVOS_M7000_Imaging_System_UG.pdf).

    Can I upgrade my EVOS M5000 Imaging System (Cat. No. AMF5000)?

    Yes, you can add different objectives, wavelength LEDs, vessel holders, OnStage Incubation (temp/humidity/CO2/hypoxic), even image analysis software (Celleste) at any time. If needed, we also offer an upgrade path to the fully automated EVOS M7000 Imaging System as well.
    Please contact Technical Support at tech.support@thermofisher.com for more information.

    How do I turn on the 'phase mode' on the EVOS M7000 Imaging System for use with my phase objectives?

    The phase mode can be activated after a phase objective has been installed and assigned in the software. A small orange triangle with letter 'B' appears in the corner of the transmitted light button. This toggle can be clicked and the orange 'B' (brightfield) should change to green 'P' (phase), activating the phase mode.

    What warranty does the EVOS M7000 Imaging System come with?

    The EVOS M7000 Imaging System comes with a standard 12-month warranty. We offer an additional 1-year extended warranty that can be purchased separately at any time (contact Technical Support at techsupport@thermofisher.com for details). We also offer EVOS M7000 Imaging System Extended Warranty Package (Cat. No. A42844) that provides the EVOS M7000 Imaging System with the standard 12-month warranty plus a 1-year extended warranty.

    During a scan routine with the EVOS M7000 Imaging System, I noticed that my fluorescent samples ended up out of focus. How can I correct this?

    First , we recommend setting the autofocus function to "autofocus" in every well/ field to correct for topographical changes across the plate. Z-Offsets specify the optimal focus position in each channel relative to the focus position in other channels. Setting the correct Z-Offsets is especially important when the fluorescent markers in different channels are in different focal planes. For best results, optimize the Z-Offsets in the Capture tab, and then carry the offsets over to the Automate tab. For detailed instructions on how to set the Z-offsets, please see pages 50-52 of the EVOS M7000 User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018326_EVOS_M7000_Imaging_System_UG.pdf).

    In the EVOS M7000 Imaging System, what are the options for displaying images? What is the file format of saved images?

    Available color options for displayed images are Grayscale (16-bit) and Pseudo-color (24-bit RGB; 8-bit per RGB channel).

    Available File format for displaying and saving images are: TIFF, PNG, and JPEG.

    I'm imaging my fluorescently stained samples on EVOS M7000/ EVOS M5000 imaging system and I can't see any color on the displayed image. How do I visualize my images in color?

    Toggle image color display provides an option to display images in pseudo-color or in grayscale in the Viewing area. By default, pseudo-color display is on. Click the Image color display button to display the sample pseudo-colored in the default emission color of the selected channels. Click again for grayscale display.

    To change the default pseudo-color, you can go to Settings Tab-> Filter cubes and adjust the pseudo-color for a selected channel to the desired color.

    The EVOS M7000 Imaging System comes with 3 individual parts (microscope base, computer and LCD monitor). Can each of these be replaced individually, if necessary?

    Yes, the EVOS M7000 Imaging System is serviceable on site by our expert EVOS engineers who will be able to replace any defective parts. All service or replacement requests will be channeled through the Technical Support team (techsupport@thermofisher.com).

    How do I setup my new EVOS M7000 Imaging System?

    The installation of the EVOS M7000 Imaging System is performed by a highly trained FAS (field application specialist) who will ensure that your system is setup correctly and functions perfectly. The FAS will also provide a hands-on training for your lab members on the day of installation, to ensure that everyone feels comfortable with setting up and using the system. Technical Support can be also reached at 541-335-0353 (in North America) to assist with any additional questions. In other regions, please dial your local Technical Support contact number.

    How do the specific parameters on an objective relate to my application and how do I select the right one for my EVOS imaging system?

    Please find a comprehensive objective selection guide in the link below. It provides detailed guidance based on sample type, vessel type and mode of imaging.
    https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/cell-imaging-systems/evos-objectives/selection-guide-evos-objectives.html


    For Research Use Only. Not for use in diagnostic procedures.