Learn More
Invitrogen™ EVOS™ M7000 Computer Upgrade
AMEP5023 contains an upgraded Dell XE4 PC and EVOS Deconvolution license (AMEP5022).
Supplier:  Invitrogen™ AMEP5023
Catalog No. AMEP5023
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The latest EVOS M7000 Imaging System is powered by an external Dell XE4 computer with Intel Core i9 processor. The Dell XE4 computer offers increased memory (128 GB DDR4 RAM) and storage (2 TB SSHD) compared to previous computer models, along with similar NVIDIA Quadro RTXA4000 with 8 GB discrete video graphics and Windows 10 21H2 LTSC operating system.
If you currently own an EVOS M7000 system with a previous model computer (Dell XE3 or Dell 3650), then purchase of this product enables an upgrade to the latest computer platform offered for the EVOS M7000 system, with the benefit of improved processor and increased memory and storage.
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
EVOS M7000 Imaging System
Dell XE4 PC with 2D deconvolution license
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.