Learn More
Invitrogen™ EVOS™ M7000 2D Deconvolution License
Catalog No. AMEP5022
Change view
Click to view available options
Product Line:
EVOS
Catalog No. Product Line
AMEP5022 EVOS
1 options
Catalog No. AMEP5022 Supplier Invitrogen™ Supplier No. AMEP5022
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AMEP5022 enables current M7000 customers having a Dell XE3, Dell 3650, or Dell XE4 to activate EVOS 2D deconvolution. Dell XE3 and 3650 computers require PC re-image by a Field Service Engineer.
The EVOS M7000 2D Deconvolution License allows users to enable 2D deconvolution on their EVOS M7000 Imaging System without having to purchase a complete third-party image analysis software suite, thus offering a significant savings. Enablement of the license requires the latest version of EVOS M7000 software or a system with a Dell XE4- model computer (upgrade to this model = Cat. No. AMEP5023).
The EVOS M7000 Imaging System is a high-performance, fully automated, inverted, multi-channel fluorescence and transmitted light imaging system. It was developed to meet today's expectations for image quality, user interface interactions, data generation speed and flexibility, and end-user value. During post-acquisition processing, one of the most commonly used techniques is deconvolution. In 2D deconvoluton, a mathematical point-spread function is applied to the image(s) as a way to increase signal:noise by reducing background.
Specifications
Product Line EVOS
For Use With (Equipment) EVOS M7000 Imaging System
For Research Use Only. Not for use in diagnostic procedures.