Learn More
Invitrogen™ EVOS™ FL Condenser Slider Block
Description
- Improves image quality
- Serves as alternative to light shield box for sample access during fluorescence imaging in dark environments
- Compatible with EVOS FL , FL Color, and FL Auto imaging systems
Specifications
Specifications
| Type | Fl Condenser Slider Block |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Condensing Optics, Hard-Coated Filters, LED |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.