Learn More
Invitrogen™ EVOS™ Condenser Slider, Diffusion
Description
- Used to improve illumination under certain conditions
- Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
- Intended for brightfield applications at 2x or 4x magnification for flat-field illumination
Diffusion condenser slider is included with EVOS™ XL Imaging System.
Specifications
Specifications
| Type | Condenser Slider |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Diffuser Slider, Pinhole Slider, Meniscus Sliders |
| Quantity | 1 each |
For Research Use Only. Not for use in diagnostic procedures.