Learn More
EVOS™ EVOS™ Condenser Slider, Meniscus A (clear liquids)
Catalog No. 12563600
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
12563600 1 Ea.
1 options
Catalog No. 12563600 Supplier EVOS™ Supplier No. AMEPMN3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Even out field of view
  • Used to improve illumination under certain conditions 
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Meniscus A slider is intended for brightfield applications at 2x magnification with low-volume samples in multi-well plates and with clear liquids
  • Meniscus B slider is intended for brightfield applications at 2x magnification with high-volume samples in multi-well plates and with semi-opaque liquids
Specifications
Type Meniscus A Condenser Slider
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Diffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity 1 Ea.
For Research Use Only. Not for use in diagnostic procedures.