Learn More
Invitrogen™ EVOS™ Condenser Slider, Green
Catalog No. 12563598
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
12563598 Each
1 options
Catalog No. 12563598 Supplier Invitrogen™ Supplier No. AMEPCFG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Adds contrast when imaging
  • Used to improve illumination under certain conditions 
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Enables direct adjustment of white light LED color temperature
  • Intended for brightfield applications at all magnifications as daylight-correction filter
Specifications
Type Condenser Slider
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Diffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity Each
For Research Use Only. Not for use in diagnostic procedures.