Learn More
Invitrogen™ 50X Oil Objective, achromat, coverslip-corrected
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Specifications
| Type | Objective Lens |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Focus Distance | 0.19 mm |
| Lens Type | Plan Achromat |
| Magnification | 50X |
| Numerical Aperture | 0.95 |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.