Learn More
Invitrogen™ EVOS™ 40X Objective, fluorite, coverslip-corrected
Catalog No. 12563587
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
12563587 1 Ea.
1 options
Catalog No. 12563587 Supplier Invitrogen™ Supplier No. AMEP4699
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For fluorescence and demanding transmitted-light applications
  • Can be used to view images with different magnification
  • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
Specifications
Type Objective Lens
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Focus Distance 0.72 mm
Lens Type Plan Fluorite
Magnification 40X
Numerical Aperture 0.75
Quantity 1 Ea.
For Research Use Only. Not for use in diagnostic procedures.