missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ 100X Oil Objective, fluorite, coverslip-corrected

Catalog No. 12563664
Change view
Click to view available options
Quantity:
1 objective
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12563664 1 objective
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12563664 Supplier Invitrogen™ Supplier No. AMEP4696

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This fluorite objective is ideal for fluorescence and demanding transmitted-light applications.

This is a coverslip-corrected objective that is optimized for imaging through No. 1.5 coverslips. The extensive choice of objectives satisfies needs across the spectrum of magnifications and optical specifications.

Additional characteristics of this EVOS objective:

  • Magnification: 100X
  • Numerical Aperture: 1.28
  • Working Distance: 0.21 mm
  • Immersion: oil

Microscope objectives may be the most important components of an optical microscope because they are responsible for primary image formation. Image quality is crucial to experimental success and a requirement for publication; EVOS objectives afford that quality across the visible spectrum to near infrared light. This performance results from years of lens manufacture perfection. EVOS objectives have the same or better numerical apertures as any other manufacturer's in the same class, and the broad selection means you have choices for your imaging requirements.

Achromat objectives are perfect for general applications, with standard correction of color and focus.

Fluorite objectives deliver excellent resolution and are made with higher numerical apertures than achromat objectives, resulting in brighter fluorescence signal and higher contrast imaging. The higher optical quality greatly reduces optical aberrations, and corrections for color and focus are at higher levels than achromat objectives. Fluorite objectives are ideally suited for fluorescence and demanding transmitted light applications, where the higher contrast make them ideally suited for color imaging.

Apochromat objectives are manufactured to the highest levels of resolution, fluorescence brightness, and contrast; chromatic aberrations are almost eliminated. They are recommended for the most demanding applications, particularly at magnifications of 60x and above. Apochromatic objectives are the best choice for the capture of color images in white light.

Brightfield is the most basic form of light microscopy and is accomplished by sample absorption of light. A higher density area in a sample will absorb more light, thus increasing contrast in those areas.

Phase contrast objectives are most useful for hard to see, translucent specimens. This method of contrast is accomplished by converting phase shifts, caused by light passing through a translucent specimen, into brightness changes (i.e., contrast).

Long working distance (LWD) objectives are optimized for use through vessels with a nominal wall thickness of 0.9-1.5 mm. This includes vessels commonly used in cell culture and cell-based assays, such as slides, cell culture dishes and flasks, microtiter well plates, etc. Coverslip-corrected objectives are optimized for use through #1.5 coverslips (thickness approximately 0.17 mm). These objectives have a higher magnification/NA ratio and provide higher resolution compared to LWD objectives.

Specifications

Type Objective Lens
Product Line EVOS
For Use With (Equipment) EVOS FL Auto 2 (AMAFD2000), EVOS M5000 (AMF5000)
Includes Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Lens Type Coverslip corrected oil immersion objective, Fluorite
Magnification 100X
Quantity 1 objective
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.