missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Evodiamine, 98%
CAS: 518-17-2 | C19H17N3O | 303.37 g/mol
$145.29 - $145.29
Chemical Identifiers
| CAS | 518-17-2 |
|---|---|
| Molecular Formula | C19H17N3O |
| Molecular Weight (g/mol) | 303.37 |
| MDL Number | MFCD06407824 |
| InChI Key | TXDUTHBFYKGSAH-SFHVURJKSA-N |
| Synonym | evodiamine, +-evodiamine, unii-c01825bvnl, evodiamine, evodia rutaecarpa, d-evodiamine, evodiamine, +, s-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, pubchem18244, indolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, 8,13,13b,14-tetrahydro-14-methyl-, 13bs, 13bs-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one |
| PubChem CID | 442088 |
| ChEBI | CHEBI:4948 |
| IUPAC Name | (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one |
| SMILES | CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC457890010
|
Thermo Scientific Chemicals
457890010 |
1 g |
Each for $145.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 518-17-2 | |
| 303.37 | |
| TXDUTHBFYKGSAH-SFHVURJKSA-N | |
| 442088 | |
| (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one |
| C19H17N3O | |
| MFCD06407824 | |
| evodiamine, +-evodiamine, unii-c01825bvnl, evodiamine, evodia rutaecarpa, d-evodiamine, evodiamine, +, s-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, pubchem18244, indolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, 8,13,13b,14-tetrahydro-14-methyl-, 13bs, 13bs-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one | |
| CHEBI:4948 | |
| CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12 |
Specifications
| 518-17-2 | |
| 97.5% min. (HPLC) | |
| C19H17N3O | |
| 1 g | |
| TXDUTHBFYKGSAH-SFHVURJKSA-N | |
| (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one | |
| 442088 | |
| 303.36 | |
| Evodiamine |
| Authentic | |
| Glass Bottle | |
| MFCD06407824 | |
| evodiamine, +-evodiamine, unii-c01825bvnl, evodiamine, evodia rutaecarpa, d-evodiamine, evodiamine, +, s-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, pubchem18244, indolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, 8,13,13b,14-tetrahydro-14-methyl-, 13bs, 13bs-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one | |
| CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12 | |
| 303.37 | |
| CHEBI:4948 | |
| 98% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
GHS Signal Word: Danger
RUO – Research Use Only