missing translation for 'onlineSavingsMsg'
Learn More

Evidence Marking Flags

Catalog No. S99537
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S99537 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99537 Supplier Customer Channels Group (Ccg) Supplier No. EVFY100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Make evidence easy to relocate or photograph.

  • Brightly colored yellow evidence flags
  • “Evidence — Do Not Touch” printed in black ink on each flag
  • 21 in. high (53.3cm)

Specifications

Color Yellow
For Use With (Application) For make evidence easy to relocate or photograph
Product Type Marking Flag