Learn More
Caplugs™ (Evergreen Scientific) Test Tubes-R-Us™ General Purpose Polypropylene Test Tubes
Precision molded from premium-grade plastics—ideal for routine laboratory analyses and procedures
Supplier:  Caplugs™ (Evergreen Scientific) 214 2301 030
 View more versions of this product
Catalog No. 22047416
Only null left
Description
Description
  • For use with high-volume clinical chemistry analzyers
  • Polypropylene tubes are more chemically resistant and are appropriate for use with organic solvents and stronger acids (except oxidizing acids) and alkalis, alcohols, chlorinated and aromatic hydrocarbons (except xylene) and ketones
  • Also available are polystyrene tubes (see cat. no. 05-555-8), which provide clarity and are appropriate for use with mild acidic (except oxidizing acids) or alkali solutions, alcohols (except higher alcohols) and salt
  • Tubes are bulk packed, non-sterile, round or conical bottom
  • Stopper caps and sterile tubes are also available
Specifications
Chemical Identifiers
Polypropylene
Non-sterile
General Purpose Test Tube
Polypropylene tubes are more chemically resistant and are appropriate for use with organic solvents and stronger acids (except oxidizing acids) and alkalis, alcohols, chlorinated and aromatic hydrocarbons (except xylene) and ketones
Not Included
Specifications
1.6 mL
Polypropylene
Non-sterile
Not Included
50 mm
Polypropylene tubes are more chemically resistant and are appropriate for use with organic solvents and stronger acids (except oxidizing acids) and alkalis, alcohols, chlorinated and aromatic hydrocarbons (except xylene) and ketones
8 mm
General Purpose Test Tube
Product Suggestions
Product Suggestions