missing translation for 'onlineSavingsMsg'
Learn More

Caplugs™ (Evergreen Scientific) Technicon™ Conical Bottom Sample Cups

Catalog No. p-3238060
Change view
Click to view available options
Volume (Metric):
2 mL
4 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
0555836 2 mL
0254472A 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0555836 Supplier Caplugs™ (Evergreen Scientific) Supplier No. 127 0090 010
Only null left

For use with Technicon Auto-Analyzers

Specifications

Product Type Sample Cup
Material Polystyrene
Volume (Metric) 2 mL
Color Natural
For Use With (Application) Automated chemistry analyzers
For Use With (Equipment) Technicon auto-analyzer
Shape Conical