missing translation for 'onlineSavingsMsg'
Learn More

Caplugs™ Evergreen Scientific Piggyback Serum Cups

Catalog No. 055524
Change view
Click to view available options
Capacity (Metric):
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
055524 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 055524 Supplier Caplugs™ Supplier No. 127 1225 010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For laboratories that use high-speed analyzers

  • Compatible with 12- and 13-mm tubes (see Evergreen Scientific Test Tubes-R-Us™ test tubes)
  • Piggyback serum cups are offered for short serum samples; constructed of polystyrene
  • Use with high-volume automate analyzers such as DuPont™ Dimension™, Boehringer Mannheim™/Hitachi™ 747, Kodak™ Ektachem™ 700 series systems, and other direct-tube sampling machines
  • To minimize variation caused by evaporation during processing, combine with Cat. No. 05-551-5 perforated flat-top caps; for short term storage, combine with non-perforted flat-top caps (Cat. No. 02-544-73A) and push-top stoppers (Cat. No. 05-300-291)
  • Cups are available in pack of 1000

Specifications

Capacity (Metric) 1 mL
Product Type Piggyback Serum Cup
Capacity (English) 0.03 oz.
Material Polystyrene
For Use With (Application) Short Serum Samples
For Use With (Equipment) High volume clinical chemistry analyzers
Capacity 1 mL