missing translation for 'onlineSavingsMsg'
Learn More

Caplugs™ (Evergreen Scientific) Disposable Funnels

Economical funnels for a variety of applications.

$103.94 - $103.94

Specifications

Diameter (Metric)Top 6 cm
Angle 60° Tapered
Includes Internal ribs
View More Specs

Products 1
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
05-555-6
View Documents
Caplugs™ (Evergreen Scientific)
2085136030
Polypropylene
Case of 100 Each for $103.94
Only null left
 
Description

Description

Choose the funnel that best meets your needs:

  • Polystyrene funnels for use with mild acidic or alkali solutions
  • Polypropylene funnels for use with organic solvents and stronger acids or alkalis

 

 

  • Funnels measure approximately 2.4 in. (60 mm) wide at the top and taper 60°
  • Internal ribs promote faster flow, external ribs help prevent airlock
  • New material specifications now include up to 50% reused plastic

 

Specifications

Specifications

6 cm
60° Tapered
Internal ribs
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'