Learn More
Carson™ EverGlade™ HD Waterproof Spotting Scope with Table-Top Tripod
Catalog No. S98154
Encompass
Change view
Click to view available options
Magnification:
15 to 45x
Catalog No. Magnification
S98154 15 to 45x
1 options
Catalog No. S98154 Supplier Carson™ Supplier No. SS560
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A powerful, compact, and lightweight high definition angled spotting scope. Nitrogen purged and O-ring sealed to make the scope both waterproof and fog-proof in extreme weather conditions.
  • A powerful, compact, and lightweight high definition angled spotting scope
  • Nitrogen purged and O-ring sealed to make the scope both waterproof and fog-proof in extreme weather environments
  • A zoom ring located on the eyepiece makes it possible to adjust the magnification between 15x and 45x power
  • The 60 mm objective lens diameter enables ideal light gathering capabilities for brighter and sharper image quality
  • BAK-4 angled prism and fully multi-coated optics provide high definition and crystal clear images
  • Twist-up eyecup designed to exclude extraneous light, with a sliding sun shade to cut down on external glare
  • Includes a table-top tripod, soft protective pouch, lens/eyepiece covers, and a lens cloth
  • Mounting base is compatible with most standard tripods, therefore the included tripod may be substituted with another of the user's choosing
  • Spotting scope can also be used without removing the protective pouch
  • Backed by Carson's limited lifetime warranty
Specifications
Lens Coating Fully Multi-coated
Product Type Spotting Scope
For Use With (Application) Astronomy, Birding, Hunting, Nature Study, and Scenic Observation
Weight (English) 2.2 lb.
Includes Table-Top Tripod, Soft Protective Pouch, Lens Cloth, and Lens/Eyepiece Covers
Magnification 15 to 45x
Model SS-560