Learn More
Biotium EVERBRITE MOUNTING MEDIUM
Supplier:  Biotium 23002
Encompass
EverBrite mounting medium is a revolutionary antifade mounting medium optimally formulated for preserving fluorescence of Biotium's CF dyes and other fluorochromes. Unlike VectaShield, EverBrite is compatible with cyanine-based fluorophores like Cy dyes. The mounting medium permits immediate imaging of slides and tissues after sealing edges of coverslips with nail polish or Biotiums CoverGrip Coverslip Sealant. After mounting, slides can be stored at 4C for a year or longer. EverBrite provides a high level of protection from photobleaching over a wide range of fluorescence wavelengths. EverBrite Mounting Medium has a refractive index of 1.46, which is well-matched to the refractive indices of coverslip glass and immersion oil (1.5). 10 mL. VectaShield is a registered trademark of Vector Laboratories. Cy Dye is a registered trademark of GE Healthcare.
Catalog No. NC0155006
Only null left