missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Evans Blue

CAS: 314-13-6 | C34H24N6Na4O14S4 | 960.79 g/mol

$105.40 - $344.23

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
Synonym C.I. 23860
PubChem CID 24832074
IUPAC Name tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1677409
View Documents
Thermo Scientific Chemicals
A1677409
10 g
Each for $105.40
Only null left
 
AAA1677418
View Documents
Thermo Scientific Chemicals
A1677418
50 g
Each for $344.23
Only null left
 
Description

Description

Evans Blue is used for estimation of blood volume and in double-labeling procedure for studying axonal branching. It acts as an inhibitor of L-glutamate and kainate receptor-mediated currents. It is also a P2X-selective purinoceptor antagonist. It is also useful in molecular biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

314-13-6
960.79
KBNIFDASRCWYGC-GXNXWABVSA-J
24832074
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
C34H24N6Na4O14S4
MFCD00004021
C.I. 23860
tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate

Specifications

314-13-6
MFCD00004021
C.I. 23860
KBNIFDASRCWYGC-GXNXWABVSA-J
tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
24832074
10 g
C34H24N6Na4O14S4
14,3905
Soluble in water.
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
960.79
960.82
Evans Blue
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H350

EINECSNumber : 206-242-5

RTECSNumber : QJ6440000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only