missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Evans Blue

Catalog No. AAA1677409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1677409 10 g
AAA1677418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1677409 Supplier Thermo Scientific Chemicals Supplier No. A1677409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 314-13-6 | C34H24N6Na4O14S4 | 960.79 g/mol

Evans Blue is used for estimation of blood volume and in double-labeling procedure for studying axonal branching. It acts as an inhibitor of L-glutamate and kainate receptor-mediated currents. It is also a P2X-selective purinoceptor antagonist. It is also useful in molecular biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00004021
InChI Key KBNIFDASRCWYGC-GXNXWABVSA-J
Synonym C.I. 23860
PubChem CID 24832074
IUPAC Name tetrasodium;4-amino-6-[[4-[4-[2-(8-amino-1-oxo-5,7-disulfonatonaphthalen-2-ylidene)hydrazinyl]-3-methylphenyl]-2-methylphenyl]hydrazinylidene]-5-oxonaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1/C=CC2=C(C=C(C(N)=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2/C=CC3=C(C=C(C(N)=C3C2=O)S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Merck Index 14,3905
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight 960.82
Chemical Name or Material Evans Blue
Hazard Category H350
Hazard Statement GHS H Statement
H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 206-242-5
RTECSNumber QJ6440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only